Dewar Shielded Vacuum Flasks, Pope Scientific
Dewar Shielded Vacuum Flask, 665 ml (22.5 oz.)
Catalog # 63422-040
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Supplier: POPE SCIENTIFIC COMPANY
Número de proveedor #: 8640-0099
Dewar Shielded Vacuum Flasks, Pope Scientific
Catalog # 63422-040
Supplier: POPE SCIENTIFIC COMPANY
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Jump to:
Product Details & Documents
These flasks are manufactured for use with liquid nitrogen (LN2) and ideal for use with liquid gases
The glass container is attached firmly to the interior of the aluminum case. Flasks are completely silvered and evacuated, ready for use.
A strong, lightweight aluminum shield encases the flask from cover to base and also serves as a stand. Accidental breakage from dropping or other impact is reduced.
Especificaciones
- Description:Dewar Shielded Vacuum Flask, 665 ml (22.5 oz.)
- Cat. No.:63422-040
- Int.Ø:70 mm (2.7")
- Capacity:0.665 L
- Overall height:241 mm (9.5")