Lab-Grade Dewar Flasks, Pope Scientific
Dewar vacuum flask, 1 L (33.8 oz.)
Catalog # 63397-000
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Supplier: POPE SCIENTIFIC COMPANY
Número de proveedor #: 8645
Lab-Grade Dewar Flasks, Pope Scientific
Catalog # 63397-000
Supplier: POPE SCIENTIFIC COMPANY
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Jump to:
Product Details & Documents
These vacuum lab-grade standard dewars are manufactured for efficient use with liquid nitrogen (LN2) and other liquid gases.
Dewar vacuum flasks are made of double-walled borosilicate glass, fully silvered, evacuated to 10-5 and covered by a protective mesh. Evacuation tips and flask bottoms are fully coated and enclosed in an aluminum base. Polyethylene lids are included with all flasks.
: Replacement lids are also available; contact your Avantor sales representative for more information.
Especificaciones
- Description:Dewar vacuum flask, 1 L (33.8 oz.)
- Cat. No.:63397-000
- Int.Ø:70 mm
- Capacity:1 L
- Int. height:302 mm
- Ext.Ø:86 mm
- Overall height:337 mm