About this item
VWR® Silicone pump tubing is extremely flexible and specially formulated for reliable performance in peristaltic pumps.
- Extremely flexible low-volatile grade platinum-cured silicone tubing
- Ideal for pharmaceutical, biomedical, laboratory, food, beverage, and fragrance applications
- Ultra-smooth interior surface for uniform flow and bacteria resistance
- Imparts no taste or odor to critical stream
Extremely flexible tubing is specially formulated to withstand repeated compression and release for consistent, reliable performance in peristaltic pumps.
Tubing has undergone extensive physical, chemical, and biological testing. Sterilizable by autoclave and gamma radiation.
Low-volatile grade, platinum-cured silicone tubing is produced in a certified ISO Class 6 (FED-STD-209E Class 1000/M4.5) cleanroom for use in critical pharmaceutical, biomedical, laboratory, food, beverage, and fragrance applications. Designed to maintain a long service life, tubing features an ultra-smooth interior surface that provides uniform fluid flow and resistance to bacteria build-up.
Specifications
- Catalog No:
- 89068-432
- 89068-440
- 89068-444
- 89068-436
- Dureza en durómetro:
- Shore A, 50
- Shore A, 50
- Shore A, 50
- Shore A, 50