HelixMark® Standard Platinum-Cured Silicone Tubing, Wacker Material, Freudenberg Medical
HelixMark® platinum-cured silicone tubing meets the needs of a broad range of applications and is universally applicable for the transfer of fluids under standard requirements.
- Fully autoclavable and sterilizable
- Ideal for use in transfer operations as well as other applications requiring non-leachable material
- Material certification in every package
- Lot traceability
HelixMark® tubing meets USP Class VI and EP 3.1.9 biocompatiblity requirements. It is manufactured in Freudenberg Medical's FDA registered medical device facility following Good Manufacturing Practices (GMPs) in accordance with The Quality Systems Requlations.
Durometer hardness: Shore A, 60.
: Materials approved in accordance with USP Class VI, ISO 10993-1, FDA 21CFR 177-2600 and EP 3.1.9 or EP 3.1.9 without appearance of solution. Certified to meet ISO 13485 quality standards.
: Tubing is packaged double-bagged in crush-resistant boxes.
Frequently Bought Together