HelixMark® Close Tolerance Platinum-Cured Silicone Tubing, Dow Corning Material, Freudenberg Medical
HelixMark® close tolerance platinum-cured silicone tubing maintains particularly tight tolerances with regard to inner diameter and wall thickness and is subjected to stringent quality assurance measures in production.
- Fully autoclavable and sterilizable
- Ideal for use in transfer operations as well as other applications requiring non-leachable material
- Material certification in every package
- Lot traceability
- Durometer hardness: Shore A, 50
HelixMark® tubing meets USP Class VI and EP 3.1.9 biocompatiblity requirements. It is manufactured in Freudenberg Medical's FDA registered medical device facility following Good Manufacturing Practices (GMPs) in accordance with The Quality Systems Requlations.
: Certified to meet ISO 13485 quality standards. Materials approved in accordance with USP Class VI, ISO 10993-1, FDA 21CFR 177-2600 and EP 3.1.9 or EP 3.1.9 without appearance of solution.
: Tubing is packaged double-bagged in crush-resistant boxes. Tubing is individually packaged in 50 foot coil.
Frequently Bought Together