Orders LinkContactUsLinkComponent
EasySat™ Bucketless Floor Mops, Contec®
EasySat™ Bucketless Floor Mops, Contec®
EasySat™ Bucketless Floor Mops, Contec®
Supplier:  CONTEC, INC. MX
Certificates
EasySat™ Bucketless Floor Mops, Contec®
EasySat™ Bucketless Floor Mops, Contec®
2 Options Available Below
Product Details & Documents
Designed specifically for cleanroom use, this mop system completely eliminates the need for bucket or wringer.
  • Hydrophilic polyester/clean ester, laser cut sealed edge foam material
  • Translucent tank allows user to see solution
  • Fully autoclavable for critical areas
  • Easy to use
  • Compatible with a wide range of solvents
System includes a 1.5 l (52 oz.) translucent solution tank that can clean up to 5,000 square feet. Tank weighs only 3.2 kg (7 lbs.) when full. A push button flow control releases cleaning solution on demand. The angled, 55.9 cm (22") mop heads clean hard-to-reach areas and can be attached and removed within seconds.
System and mop heads are fully autoclavable up to 121 °C (250 °F) for 30 minutes. Mop heads come in regular and textured styles and are also available gamma irradiated.
It features lasercut sealed-edge VertiKlean® mop head material, which is a lightweight polyester fabric laminated to clean ester foam for excellent abrasion resistance.
Ordering information: Mop system and mop heads must be purchased separately.
Product Family Options
Product Information
  • Dimensions
    Material
    Packed
    Availability
    Price
  • 33501-534
    EasySat™ Bucketless floor mop system
    EasySat™ Bucketless floor mop system
    55×46×12 cm (21.5×18.25×4.75")
    -
    1/case
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $10,166.85
    $10,166.85
    Each (1UN)
       $10,166.85
    Supplier Number: BLMOP
  • 33501-540
    EasySat™ Textured mop head
    EasySat™ Textured mop head
    55×12×46 cm (21.5×4.75×18.25")
    Textured Knitted Polyester/Foam
    4/bag, 8 bags/case
    Discontinued
    Supplier Number: BLVKMOP
  • 33501-536
    EasySat™ Mop head
    EasySat™ Mop head
    55×12×46 cm (21.5×4.75×18.25")
    Knitted Polyester/Foam
    4/bag, 8 bags/case
    Log in for availability
    Log in for availability
    Case (32UN)
    Case (32UN) - $10,438.90
    $10,438.90
    Case (32UN)
       $10,438.90
    Supplier Number: BLVKMOP