Orders LinkContactUsLinkComponent
AlphaMop™ Polyester Mop
AlphaMop™ Polyester Mop
Supplier:   Texwipe
AlphaMop™ Polyester Mop
Supplier:  Texwipe
Certificates
AlphaMop™ Polyester Mop
AlphaMop™ Polyester Mop
5 Options Available Below
Product Details & Documents
Ideal for cleaning walls, floors, and other flat surfaces in critical environments.
AlphaMop™ covers TWTX7026 are pre-wetted with 6% isopropyl alcohol. The case purchase includes two foam pads. Replacement pack TW-TX7118 contains 150 polyester heads and 6 foam pads.
The changeable, laundered polyester head fits over a flat foam swivel pad assembly attached to a fiberglass handle. Replaceable foam pad ensures the polyester head conforms to textured floors and other surfaces while resisting corrosion and abrasion. Lightweight mop maintains ultralow generation of particles and extractables. Bag-within-a-Bag® packaged.
Product Family Options
Información del producto
  • Packed
    Availability
    Price
  • TWTX7108
    AlphaMop™ with head, pad, and handle
    AlphaMop™ with head, pad, and handle
    10 covers and 1 pad/bag; 12 bags/case
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $4,140.00
    $4,140.00
    Each (1UN)
       $4,140.00
    Supplier Number: TX7108
Replacement Parts
Información del producto
  • Packed
    Availability
    Price
  • TW-TX7118
    AlphaMop™ polyester heads and foam pads
    AlphaMop™ polyester heads and foam pads
    10 covers and 1 pad/bag; 12 bags/case
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (150UN)
    Case (150UN) - $23,440.00
    $23,440.00
    Case (150UN)
       $23,440.00
    Supplier Number: TX7118
  • 10127-118
    Autoclavable replacement head
    Autoclavable replacement head
    10 covers and 1 pad/bag; 12 bags/case
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $4,330.00
    $4,330.00
    Each (1UN)
       $4,330.00
    Supplier Number: TX7108AH
  • 10127-132
    Replacement handle, 1.5 m (60")
    Replacement handle, 1.5 m (60")
    10 covers and 1 pad/bag; 12 bags/case
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $2,165.24
    $2,165.24
    Each (1UN)
       $2,165.24
    Supplier Number: TX7123
  • 10127-120
    Replacement head
    Replacement head
    10 covers and 1 pad/bag; 12 bags/case
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $2,610.00
    $2,610.00
    Each (1UN)
       $2,610.00
    Supplier Number: TX7108H