VWR® Dippers, PTFE
Dippers feature a 60 cm (235/8") long steel shaft encapsulated in PTFE for rigidity.
- Steel fabrication
- Encapsulated in PTFE for rigidity
- Sample most aggressive media confidently
Suitable for sampling the most aggressive media.
: Dippers are supplied with one shaft; additional shafts (89026-222) are available to extend length.
Dippers feature a 60 cm (235/8") long steel shaft encapsulated in PTFE for rigidity.
- Steel fabrication
- Encapsulated in PTFE for rigidity
- Sample most aggressive media confidently
Suitable for sampling the most aggressive media.
: Dippers are supplied with one shaft; additional shafts (89026-222) are available to extend length.
Product Family Options
- DescriptionØCapacity