VWR® Thin Sleeves, PTFE
These PTFE sleeves withstand low to medium pressure and are inserted between standard ground glass joints to eliminate the use of grease and prevent sticking.
- Chemically resistant
- Inert
- PTFE fabrication
Sleeves are inert, have a wall thickness of about 0.05mm, and are usable to 280°C (536°F).
These PTFE sleeves withstand low to medium pressure and are inserted between standard ground glass joints to eliminate the use of grease and prevent sticking.
- Chemically resistant
- Inert
- PTFE fabrication
Sleeves are inert, have a wall thickness of about 0.05mm, and are usable to 280°C (536°F).