To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Descripción:Mini Tray (Tray Only)
- Referencia:30109-502
- La x An x Al:165x229x51 mm (6.5x9x2")
- Nº de proveedor:JAC10
Specifications
About this item
Polystyrene trays include components to assist in efficient blood collection
All replacement components (except HDPE containers) are constructed of translucent white autoclavable polypropylene. The utility boxes contain one removable divider and are useful for carrying a variety of small items. The gauze box has one removable divider that creates three compartments total: one large 11.7Wx11.7Dcm (45/8x45/8") compartment and two smaller 6.4Wx7Dcm (21/2x23/4") compartments. The packet/tube holder is ideal for BD Vacutainer® tubes, packets, winged infusion sets, and syringes. It features three removable dividers to provide up to six compartments. The 237mL (8oz.) HDPE containers feature snap-on lids and have excellent resistance to acids, alcohols, and bases.