Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Especificaciones
- Color:White
- Referencia:14230-492
- Empaquetado:Bulk
- Units:Pack of 250/Case of 1000
- Nº de proveedor:M503-2
Especificaciones
Acerca de este artículo
Molded from a special high density polymer, these disposable cassettes keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents. Cassettes have six numbered compartments measuring 7x12mm (5/16x1/2"). The cover and base have over 3000 square openings of 0.26mm to maximize fluid exchange and ensure proper drainage. The mesh ensures efficient fluid exchange and drainage. The one-piece integral lid eliminates the need for separate steel lids. They can be opened and closed as often as necessary and they always relock securely without danger of specimen loss. Large labeling areas are located on three sides of the cassettes for convenience. Anterior writing area is at a 45° angle to make the cassette more suitable and for use with certain types of cassette labeling instruments. Available in 11 noncytotoxic, nonmetallic colors.
- With six separate compartments
- Unobstructed anterior writing area at a 45° angle
- No biopsy pad necessary
- Compatible with most cassette printers
- Lid is pre-attached to base when bulk
- Lid is seperate when in sleeve
This model can hold up to six tissue specimens, each one placed in its own 7×12 mm (¹/₄×⁷/₁₆") compartment, numbered from 1 to 6. Cover and base have over 2000 square openings to maximize fluid exchange and ensure proper drainage. Approximately 170 holes (each having a diameter of 0.26 mm) per compartment.