Orders LinkContactUsLinkComponent
VWR® Specific Gravity Precision Hydrometers
VWR® Specific Gravity Precision Hydrometers
VWR® Specific Gravity Precision Hydrometers
Supplier:  VWR International
Certificates
VWR® Specific Gravity Precision Hydrometers
13 Options Available Below
Product Details & Documents
Feature 0.0005 divisions, 325mm length and metal ballast. Hydrometers include individual serial numbers and multi-language (English, French, German, Italian, Portuguese, Spanish) Statement of Accuracy indicating accuracy traceable to NIST and DKD/PTB.
  • 325 mm (12.8”) long
  • Lead free ballast and glass
  • Individually serialized
  • Temperature standardization: 60°F (15°C)
  • Hand inspected by our U.S. quality team
Specific gravity hydrometers are the most widely used hydrometer, common in industries such as food and beverage, chemical, pharmacology, transportation, refrigeration and more.
Packaging: Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST.
Services: VWR CATALYST™ provides convenient and efficient re-calibration services for your hydrometers with an industry leading 5-day average turn-around time from receipt of your instrument. All the service will be performed by an ISO17025 certified lab. Recalibration, certification and shipping fees are all included in the catalog number cost. Simply order the stated catalog number based on how many calibration points are desired. You will receive a protective box to ship your instrument to Avantor's certified calibration laboratory along with a work order form to specify your calibration points and prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defects under a microscope and the performance of your instrument will be compared to certified standards and corrections to one-tenth of the smallest division recorded on your signed calibration report. Your calibration report and instrument are carefully repacked returned to you with prepaid standard ground shipping. We also provide expedited service options available upon request.
Documents
Specifications for Product Family
Especificaciones
  • Catalog No:
  • 34627-118
  • 34627-173
  • 34627-174
  • 34627-231
  • 34627-253
  • 34627-297
  • 34627-311
  • 34627-355
  • 34627-388
  • 34627-413
  • 34627-446
  • 34627-560
  • 34627-628
  • Accuracy:
  • 0.001
  • 0.001
  • 0.001
  • 0.001
  • 0.001
  • 0.001
  • 0.001
  • 0.001
  • 0.001
  • 0.001
  • 0.001
  • 0.001
  • 0.001
  • Length:
  • 325 mm
  • 325 mm
  • 325 mm
  • 325 mm
  • 325 mm
  • 325 mm
  • 325 mm
  • 325 mm
  • 325 mm
  • 325 mm
  • 325 mm
  • 325 mm
  • 325 mm
Especificaciones
Product Family Options
Hydrometers
Información del producto
  • Divisions
    Range
    Availability
    Price
  • 34627-118
    Precision Specific Gravity Hydrometers
    Precision Specific Gravity Hydrometers
    0.0005
    0.760 - 0.830
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $729.82
    $729.82
    Each (1UN)
       $729.82
    Especificaciones:
    More
    Specifications
    Cat. No.
    34627-118
    Accuracy
    0.001
    Length
    325 mm
  • 34627-173
    Precision Specific Gravity Hydrometers
    Precision Specific Gravity Hydrometers
    0.0005
    0.880 - 0.950
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $724.99
    $724.99
    Each (1UN)
       $724.99
    Especificaciones:
    More
    Specifications
    Cat. No.
    34627-173
    Accuracy
    0.001
    Length
    325 mm
  • 34627-174
    Precision Specific Gravity Hydrometers
    Precision Specific Gravity Hydrometers
    0.0005
    0.940 - 1.010
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $721.63
    $721.63
    Each (1UN)
       $721.63
    Especificaciones:
    More
    Specifications
    Cat. No.
    34627-174
    Accuracy
    0.001
    Length
    325 mm
  • 34627-231
    Precision Specific Gravity Hydrometers
    Precision Specific Gravity Hydrometers
    0.0005
    1.000 - 1.070
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $763.36
    $763.36
    Each (1UN)
       $763.36
    Especificaciones:
    More
    Specifications
    Cat. No.
    34627-231
    Accuracy
    0.001
    Length
    325 mm
  • 34627-253
    Precision Specific Gravity Hydrometers
    Precision Specific Gravity Hydrometers
    0.0005
    1.060 - 1.130
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $721.41
    $721.41
    Each (1UN)
       $721.41
    Especificaciones:
    More
    Specifications
    Cat. No.
    34627-253
    Accuracy
    0.001
    Length
    325 mm
  • 34627-297
    Precision Specific Gravity Hydrometers
    Precision Specific Gravity Hydrometers
    0.0005
    1.120 - 1.190
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $721.78
    $721.78
    Each (1UN)
       $721.78
    Especificaciones:
    More
    Specifications
    Cat. No.
    34627-297
    Accuracy
    0.001
    Length
    325 mm
  • 34627-311
    Precision Specific Gravity Hydrometers
    Precision Specific Gravity Hydrometers
    0.0005
    1.180 - 1.250
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $721.78
    $721.78
    Each (1UN)
       $721.78
    Especificaciones:
    More
    Specifications
    Cat. No.
    34627-311
    Accuracy
    0.001
    Length
    325 mm
  • 34627-355
    Precision Specific Gravity Hydrometers
    Precision Specific Gravity Hydrometers
    0.0005
    1.240 - 1.310
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $725.12
    $725.12
    Each (1UN)
       $725.12
    Especificaciones:
    More
    Specifications
    Cat. No.
    34627-355
    Accuracy
    0.001
    Length
    325 mm
  • 34627-388
    Precision Specific Gravity Hydrometers
    Precision Specific Gravity Hydrometers
    0.0005
    1.300 - 1.370
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $721.63
    $721.63
    Each (1UN)
       $721.63
    Especificaciones:
    More
    Specifications
    Cat. No.
    34627-388
    Accuracy
    0.001
    Length
    325 mm
  • 34627-413
    Precision Specific Gravity Hydrometers
    Precision Specific Gravity Hydrometers
    0.0005
    1.360 - 1.430
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $715.63
    $715.63
    Each (1UN)
       $715.63
    Especificaciones:
    More
    Specifications
    Cat. No.
    34627-413
    Accuracy
    0.001
    Length
    325 mm
  • 34627-446
    Precision Specific Gravity Hydrometers
    Precision Specific Gravity Hydrometers
    0.0005
    1.420 - 1.490
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $799.94
    $799.94
    Each (1UN)
       $799.94
    Especificaciones:
    More
    Specifications
    Cat. No.
    34627-446
    Accuracy
    0.001
    Length
    325 mm
  • 34627-560
    Precision Specific Gravity Hydrometers
    Precision Specific Gravity Hydrometers
    0.0005
    1.660 - 1.730
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $880.01
    $880.01
    Each (1UN)
       $880.01
    Especificaciones:
    More
    Specifications
    Cat. No.
    34627-560
    Accuracy
    0.001
    Length
    325 mm
  • 34627-628
    Precision Specific Gravity Hydrometers
    Precision Specific Gravity Hydrometers
    0.0005
    1.780 - 1.850
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $881.54
    $881.54
    Each (1UN)
       $881.54
    Especificaciones:
    More
    Specifications
    Cat. No.
    34627-628
    Accuracy
    0.001
    Length
    325 mm