J.T.Baker® BAKERBOND®, SPE Columns
: Avantor
Disposable polypropylene columns are prepacked with selective reversed phase, polar, and ion exchange organosilanes bonded to 40 µm, 60 Å, and 275 Å silica gel absorbents and size exclusion gels.
- Silica based
- Processing by vacuum or positive pressure
The highly surface-active sorbents, contained by two 20 µm polyethylene frits, provide high capacity. This configuration allows extraction of compounds from sample solution volumes of a few hundred microliters to several milliliters.
- Catalog No:
- JT70867
- JT72137
- Carga de carbono:
- —
- —
Frequently Bought Together