Orders LinkContactUsLinkComponent
VWR® Ladle, PTFE
VWR® Ladle, PTFE
VWR® ladle, PTFE, 10 ml, 170 mm
Catalog # 89026-192
Supplier:   VWR International
VWR® Ladle, PTFE
Catalog # 89026-192
Supplier:  VWR International
VWR® Ladle, PTFE
VWR® Ladle, PTFE
VWR® ladle, PTFE, 10 ml, 170 mm
Price
$1,756.25
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
This PTFE molded sampling ladle is useful for transferring hot or cold reactive substances.
  • Chemically resistant
  • Inert
  • PTFE fabrication
Especificaciones
  • Capacity:
    10 ml
  • Description:
    VWR® ladle, PTFE
  • Handle Length:
    170 mm