Orders LinkContactUsLinkComponent
VWR® Vacuum Bulb
VWR® Vacuum Bulb
Catalog # 82024-578
Supplier:   VWR International
VWR® Vacuum Bulb
Catalog # 82024-578
Supplier:  VWR International
VWR® Vacuum Bulb
VWR® Vacuum Bulb
VWR® Vacuum Bulb
Price
$275.77
Log in Para disponibilidad
Each (1UN) Case de 100 (1UN)
Product Details & Documents
Designed for use in gas sampling or for pumping gas through apparatus.
  • Heavy-walled design
  • Ideal for use in gas sampling
  • Supplied independent of tubing
Heavy-walled bulbs feature two rubber valves with PVC connections. Bulb capacity is 85 mL. Not supplied with tubing.
Especificaciones
  • Description:
    VWR® Vacuum Bulb