Transfertube® Disposable Harvesters, Spectrum® Laboratories
Catálogo # 52950-159
Proveedor: REPLIGEN CORPORATION
Small Business Enterprise
Especificaciones
- Descripción:Harvesters
- Referencia:52950-159
- Nº de proveedor:190195P
Especificaciones
Acerca de este artículo
Designed for quick and easy, one-hand transfer of agar plugs and the harvesting of microbial colonies
Just insert harvester, pull plunger, transfer, and depress plunger to release. Also recommended for taking liquid samples. Comes as a complete unit that does not require a rubber bulb. Holds approximately 4mL of liquid. Autoclavable. Made from polypropylene.