Orders LinkContactUsLinkComponent
Drum Thieves, Koehler
Drum Thieves, Koehler
Plated Brass, 710 ml (24 oz.), 32×1020 mm (1.25×40")
Catalog # 52933-312
Número de proveedor #:  K27400
Drum Thieves, Koehler
Catalog # 52933-312
Drum Thieves, Koehler
Drum Thieves, Koehler
Plated Brass, 710 ml (24 oz.), 32×1020 mm (1.25×40")
Price
$26,587.33
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Sampling tubes are suitable for taking bottom or other-level samples from barrels and drums.
Shipping weight: 2.7 kg (6 lbs.)
Especificaciones
  • Capacity:
    710 ml (24 oz.)
  • Description:
    Plated Brass
  • O.D.×L:
    32×1020 mm (1.25×40")