Orders LinkContactUsLinkComponent
VWR® Soil Analysis ASTM Hydrometers
VWR® Soil Analysis ASTM Hydrometers
VWR® Soil Analysis ASTM Hydrometers
VWR® Soil Analysis ASTM Hydrometers
VWR® Soil Analysis ASTM Hydrometers
For determining the specific gravity of soil. Conforms to ASTM specifications. Hydrometers are individually serialized and feature a metal ballast. Hydrometers include multi-language (English, French, German, Italian, Portuguese and Spanish) Statement of Accuracy indicating accuracy traceable to NIST and DKD/PTB. Tested and calibrated in an exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST and DKD/PTB. Temperature of standardization 68F/68F.
  • Lead free ballast and glass
  • Individually serialized
  • Temperature standardization: 68 /68 °F
  • Hand inspected by our U.S. quality team
Certifications: Traceable to NIST.
Packaging: Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST.
Caution: Hydrometers should be stored in a clean, dry, safe place, and should not be subjected to extreme temperatures.
Services: VWR CATALYST™ provides convenient and efficient re-calibration services for your hydrometers with an industry leading 5-day average turn-around time from receipt of your instrument. All the service will be performed by an ISO17025 certified lab. Recalibration, certification and shipping fees are all included in the catalog number cost. Simply order the stated catalog number based on how many calibration points are desired. You will receive a protective box to ship your instrument to Avantor's certified calibration laboratory along with a work order form to specify your calibration points and prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defects under a microscope and the performance of your instrument will be compared to certified standards and corrections to one-tenth of the smallest division recorded on your signed calibration report. Your calibration report and instrument are carefully repacked returned to you with prepaid standard ground shipping. We also provide expedited service options available upon request.
Documents
Product Family Options
Hydrometers
  • Description
    Range
    Accuracy
    Length
    Graduations
    ASTM No.
  • Soil Analysis Hydrometer
    SG -5/60 g/L
    ±1 g/L
    280 mm
    1.0
    152H
    Each (1UN)
    Each (1UN) - $1,018.50
    $1,018.50
    Each (1UN)
       $1,018.50
  • Soil Analysis Hydrometer
    SG 0.995/1.038
    ±0.001
    280 mm
    0.001
    151H
    Each (1UN)
    Each (1UN) - $1,204.50
    $1,204.50
    Case de 12 (1UN)
    Case de 12 (1UN) - $1,312.90
    $1,312.90
    Each (1UN)
       $1,204.50
    Case de 12 (1UN)
       $1,312.90