VWR® Sterile Softside Goggles
Product Details & Documents
Softside indirect cap safety goggles feature a green tinted frame with an indirect cap-vented frame for exceptional eye protection and splash resistance with a fog-free lens.
Green tinted frame with fog-free lens
- Indirect cap-vented frame provides splash resistance
- Silicone rubber elastic band
- Soft plastic construction for a comfortable fit and good peripheral vision
These economically priced safety goggles feature a soft construction that delivers a comfortable fit and allows for good peripheral vision. Indirect vented goggles are sterilized in the United States by Steris Isomedix Services using an ethylene oxide process.
Residuals (over eight-hour exposure period), Ethylene Chlorohydrin: 0.0429 mg
Residuals (over eight-hour exposure period), Ethylene Glycol: 0.1035 mg
Residuals (over eight-hour exposure period), Ethylene Oxide: <0.200 mg
Packaging: Each pair is individually packed in a Tyvek pouch.
Documents
Certificates
Buscar por número de loteEspecificaciones
- Frame Color:Green
- Description:Indirect ventilation goggle, sterile
- Lens Coating:Anti-fog
- Lens Tint:Clear
- Lens Material:PC