Rubber Work Overshoes, Tingley
Acerca de este artículo
100% waterproof overshoes made from a injection-molded, seamless rubber construction and a deep cleated outsole, which releases debris and provide good slip resistance. The boots stretch easily to fit even over the bulkiest workshoes.
- Reinforced heel and toe
- Excellent tensile strength
- Heavy-duty heel kick-off cleat for easy no-hands removal
- Molded-in button for secure closure
- Material will not crack or stiffen in cold weather
- Unlined for easy cleaning and decontamination
- Additional material in the heel, toe, and sole provide longer wear; less material in the upper reduces weight and leg fatigue
Designed to resist acids, alcohols, bases, and diluted water solutions of most water-soluble chemicals.
Color: Black
Height: 25.4 cm (10")
Caution: Not recommended for constant contact with petroleum and oil-based products.
3 Options Available Below
Product Details & Documents
100% waterproof overshoes made from a injection-molded, seamless rubber construction and a deep cleated outsole, which releases debris and provide good slip resistance. The boots stretch easily to fit even over the bulkiest workshoes.
- Reinforced heel and toe
- Excellent tensile strength
- Heavy-duty heel kick-off cleat for easy no-hands removal
- Molded-in button for secure closure
- Material will not crack or stiffen in cold weather
- Unlined for easy cleaning and decontamination
- Additional material in the heel, toe, and sole provide longer wear; less material in the upper reduces weight and leg fatigue
Designed to resist acids, alcohols, bases, and diluted water solutions of most water-soluble chemicals.
Color: Black
Height: 25.4 cm (10")
Caution: Not recommended for constant contact with petroleum and oil-based products.
Product Family Options
Información del producto
- Shoe sizeColorAvailabilityPrice
- SBlackEach (1UN)$1,130.35Case de 12 (1UN)$1,232.09Supplier Number: 1400.SM.01
- MBlackEach (1UN)$1,130.35Case de 12 (1UN)$1,232.09Supplier Number: 1400.MD.01
- LBlackEach (1UN)$1,130.35Case de 12 (1UN)$1,232.09Supplier Number: 1400.LG.01