ContactUsLinkComponent
VWR® SPE Glass Column Processors
VWR® SPE Glass Column Processors
Catalog # JT6998-0
MOC:  Non-RegulatedCertificates
undefined
VWR® SPE Glass Column Processors
Catalog # JT6998-0
Supplier Number:  6998-00
MOC:  Non-RegulatedCertificates
Specifications
  • Descripción:
    SPE-12G column processor
  • Referencia:
    JT6998-0
  • Nº de proveedor:
    6998-00
Specifications
About this item
These glass column processors are specially designed glass vacuum chambers that allow multiple samples to be run simultaneously. The glass chamber allows unobstructed viewing of samples at all times.
  • Run multiple samples simultaneously
  • Unobstructed viewing of samples
  • Specially-designed, glass vacuum chambers
The SPE-12G processor allows up to 12 samples to be run simultaneously and includes 12 Luer-type fittings, 12 individual flow control stopcocks, 12 polypropylene collection needles, a vacuum gauge/controller, and a rack set that accommodates a variety of glassware.
The SPE-24G processor allows up to 24 samples to be processed in 20 to 30 minutes and includes 24 Luer-type fittings, 24 individual flow control stopcocks, 24 polypropylene collection needles, a vacuum gauge/controller, and a rack set that accommodates a variety of glassware.
Ordering information: Both units are supplied with a vacuum manifold, a sample collection rack with three support posts, four adjustable height shelves with support clips, and an instruction manual.