Nunclon™ Delta 96-Well MicroWell™ Plates, Sterile, Thermo Scientific
These 96-well, polystyrene plates are ideal for cell culture, cloning, viral titration, and cell fusion
The Nunclon™Δ treated surface ensures optimal cell growth and attachment. Plates fit standard automated handling equipment. Plate edges and lids are designed to minimize evaporation. Lids are shaped to facilitate stacking. Raised well rims reduce cross-contamination. White plates provide maximum reflection, minimum autofluorescence and autoluminescence. Black plates provide minimum background and back light scatter in fluorescence assays. Plates also feature alphanumeric well identification and excellent optical quality. Radiation sterilized.
: For lids and sealing tapes, see the Nunc® Microplate Lids, Thermo Scientific series.
- Catalog No:
- 25382-341
- 21993-954
- 25382-342
- Volumen de trabajo:
- 50 - 250 µl
- 50 - 250 µl
- 50 - 250 µl
Frequently Bought Together