ContactUsLinkComponent
Difco™ L-Arabinose/Dextrose/Cellobiose, BD Biosciences
Difco™ L-Arabinose/Dextrose/Cellobiose, BD Biosciences
Catalog # 90000-908
CAS Number:  
undefined
Difco™ L-Arabinose/Dextrose/Cellobiose, BD Biosciences
Catalog # 90000-908
Supplier Number:  215530
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Descripción:
    Dextrose
  • Tamaño:
    500 g
  • Referencia:
    90000-908
  • Nº de proveedor:
    215530
Specifications
About this item
Media formulations in dehydrated form is for the convenience of the laboratorian and bioprocessing specialist.
  • Consistent, high-quality performance
  • Conveniently dehydrated
The individual ingredients and the final dehydrated product are carefully quality-controlled to assure consistent, high quality performance and obviate the need for media to be prepared in the laboratory from raw materials. Dehydrated culture media may be offered as 100 g and 500 g bottles, 2 kg and 10 kg containers.
Glucose, D (+), anhydrous, for incorporation into microbiological culture media as a source of energy for bacteria and for the determination of fermentation reactions. It is usually employed at a concentration of 0.5 to 1.0% in fermentation media.