- Color del marco:Patriot RWB
- Descripción:Uvex Astrospec 3000® eyewear with Duoflex™ temples
- Referencia:33002-049
- Recubrimiento de lentes:Scratch-resistant/hardcoat (Ultra-dura®)
- Tinte de las lentes:Clear
- Nº de proveedor:S1169
Unique shape and style provide excellent eye protection in a variety of frames and lenses. Wide viewing area provides good visibility and protection from flying particles. Glasses feature a nylon frame and lens with molded-in side shields and brow guard. Four-stop temple adjustment (130–150mm) provides a face-hugging fit. Optically correct, one-piece polycarbonate lens is lightweight and impact-resistant. Lens angle adjusts for a comfortable fit. Temples will not scratch the lens when folded.
- Wide viewing area provides good visibility and protection from flying particles
- Nylon frame and lens with molded-in side shields and brow guard
- Four-stop temple adjustment (130 to 150 mm) provides a face-hugging fit
- Optically correct, one-piece polycarbonate lens is lightweight and impact-resistant
- Lens angle adjusts for a comfortable fit
- Temples will not scratch the lens when folded
Clear lenses are for most indoor applications. Gray and silver mirror lenses minimize eye fatigue due to outdoor sunlight and glare. Amber lenses are useful for low-light applications where enhanced contrast is needed. SCT-Low IR lenses are ideal for when peripheral IR protection is required. The Patriot® RWB frame is a red, white, and blue design with miniature U.S. flags at temples and “United We Stand” inscribed on bottom right lens outside field of vision.
Eyewear meets or exceeds ANSI Z87+ and CSA Z94.3 standards.