ContactUsLinkComponent
SP Bel-Art Techni-Dome® Vacuum Desiccator, Bel-Art Products, a part of SP
SP Bel-Art Techni-Dome® Vacuum Desiccator, Bel-Art Products, a part of SP
Catalog # 89029-710
undefined
SP Bel-Art Techni-Dome® Vacuum Desiccator, Bel-Art Products, a part of SP
Catalog # 89029-710
Supplier Number:  F42029-0000
Specifications
  • Volumen:
    2.3 cu. ft (internal)
  • Descripción:
    Techni-Dome® Vacuum Desiccator (1 gas port)
  • Referencia:
    89029-710
  • An x Al:
    60×60 cm (22 x 22")
  • Nº de proveedor:
    420290000
Specifications
About this item
This domed desiccator provides a roomy interior for large, moisture-sensitive materials as well as portability and ease of use.
  • For use with moisture-sensitive materials
  • Clear polycarbonate construction
  • Unobsctructed view of contents
  • Resistant against chemicals and thermal stress
Tapered stopcock accepts ¼" to ⅜” I.D tubing and has an easy turning PTFE valve which allows for the gentle entrance of air and minimal disturbance to the inside chamber and its contents.
Used to degas liquids, porous solids, or silicon molds to eliminate air bubbles, it has a spacious 65 liter (4000 cu. in.) internal volume that can hold items up to 45 cm (18”) tall or wide, including racks, trays and instruments. It holds a full vacuum (29” Hg) at room temperature for 24 hours.
Laboratory grade polycarbonate provides a clear, unobstructed view of contents and is resistant to most common chemicals and thermal stress. Side handles provide easy portability and can be used to affix a lock or tamper-evident seals. The unit includes two removable polypropylene shelves; 36.5 cm (14.5”) and 22.5 cm (9”). U.S. Patents: 7,594,342 and D556,895 S.
Ordering information: Gas ports can be closed to use desiccator with standard desiccant cartridges; contact your Avantor representative for more information.