ContactUsLinkComponent
Uvex Ultra-spec® Protective Eyewear, Honeywell Safety
Uvex Ultra-spec® Protective Eyewear, Honeywell Safety
Catálogo # 13500-857
undefined
Uvex Ultra-spec® Protective Eyewear, Honeywell Safety
Catálogo # 13500-857
Número de proveedor:  S0112C
Especificaciones
  • Descripción:
    Ultra-spec 2001 OTG glasses
  • Referencia:
    13500-857
  • Recubrimiento de lentes:
    Uvextreme®
  • Tinte de las lentes:
    Clear
  • Nº de proveedor:
    S0112C
Especificaciones
Acerca de este artículo
Economical glasses feature a 100% polycarbonate wraparound unilens design.
  • Features 100% polycarbonate wraparound unilens design
  • Side shields add extra protection without compromising visibility
  • Clear lenses and frames
  • The Ultra-dura® lens coating provides superior scratch and chemical resistance
  • Uvextreme® lens coating provides anti-fog, anti-scratch, anti-static, and ultaviolet protection
Uvex Ultra-spec® 2000 glasses feature an adjustable temple length. Uvex Ultra-spec® 2001 OTG glasses fit over prescription lenses and feature adjustable Duoflex™ temples with soft, behind-the-ear comfort cushions. Clear lenses and frames.
The Ultra-dura® lens coating provides superior scratch and chemical resistance. The Uvextreme® lens coating provides antifog, anti-scratch, antistatic, and anti-UV protection.
Side shields add extra protection and provide optimal coverage without compromising visibility.
Certificaciones: Meet ANSI Z87+ and CSA Z94.3 specifications.