ContactUsLinkComponent
Ultra C18 (USP L1) 3 and 5 µm Columns, Restek
Ultra C18 (USP L1) 3 and 5 µm Columns, Restek
Catálogo # 89047-722
Supplier:  RESTEK CORP
undefined
Ultra C18 (USP L1) 3 and 5 µm Columns, Restek
Catálogo # 89047-722
Supplier:  RESTEK CORP
Supplier Number:  9174335
Specifications
  • Formato de columna:
    Analytical
  • Ø int.:
    4.60 mm
  • Tamaño de partícula:
    3 µm
  • Tamaño de poro:
    100 Å
  • Longitud:
    30 mm
  • Rango de pH:
    2.5 to 7.5
  • Temperature limit:
    80°C (176°F)
  • Carga de carbono:
    20%
  • Descripción:
    89047-722
  • Referencia:
    89047-722
  • Packing Material:
    Silica
  • Phase:
    C18
  • Nº de proveedor:
    9174335
Specifications
About this item
The ideal column for carbonyls, fatty acids, glycerides, anilines, fat-soluble vitamins, phthalates, barbiturates, steroids, PTH amino acids, and other acids.
  • Excellent peak shape across a range of compounds
  • Suited for general-purpose reversed phase applications
  • Fully endcapped
The retentive, high-purity packing exhibits excellent peak shape for a wide range of compounds. Columns are best suited for general-purpose reversed phase applications.