ContactUsLinkComponent
VWR® Lens Cleaning Tissues
VWR® Lens Cleaning Tissues
Catalog # 52846-001
Supplier:  VWR International
Product available on GSA Advantage®
undefined
VWR® Lens Cleaning Tissues
Catalog # 52846-001
Supplier:  VWR International
Product available on GSA Advantage®
Specifications
  • Dimensiones:
    10.1 x 15.2 cm (4 x 6")
  • Referencia:
    52846-001
  • Empaquetado:
    50 Sheets/Book, 12 Books/Pack
Specifications
About this item
Dust-free lens tissues are ideal for use with liquid cleaners.
  • High wet strength
Clean safety glasses and goggles, microscope lenses, and more with no scratching.