Eisco Gold Leaves for Electroscope
Gold leaves, pack of 6, 1.25×0.5"
Catalog # 470219-524
Supplier: EISCO SCIENTIFIC LLC
Número de proveedor #: PH0914
Jump to:
Product Details & Documents
Enhance your electroscope with these fine gold leaves.
- Pack of 6 gold leaves
- Replacements for gold leaves in electroscope
- Leaf Dimensions: 1.25" long and 0.5" wide
- Provided in plastic case
The gold leaves are spares for use in an electroscope in the event that one misplaces or damages the gold leaves used with the device (pack of 6 gold leaves). The electroscope is a tool that is used to study basic electrostatic principles.
Especificaciones
- Description:Gold leaves, pack of 6
- L×W:1.25×0.5"