ContactUsLinkComponent
VersaQuat™ Disinfectant Cleaner
VersaQuat™ Disinfectant Cleaner
Catalog # 77530-432
Supplier:  BERKSHIRE CORP
New Product
undefined
VersaQuat™ Disinfectant Cleaner
Catalog # 77530-432
Supplier:  BERKSHIRE CORP
Supplier Number:  VQUAT4G
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Descripción:
    VersaQuat™ disinfectant and cleaner, Double bagged
  • Referencia:
    77530-432
  • Empaquetado:
    Trigger spray bottle
  • Volumen:
    4 gal./512 oz.
Specifications
About this item
VersaQuat™ is an EPA-registered, RTU (ready-to-use) one-step disinfectant and cleaner which utilizes well-established and accepted quaternary ammonium chemistry with acceptable residue and safety profiles. Made in the USA with no preservatives or fragrances, Berkshire’s VersaQuat™ is a powerful and safer disinfectant alternative for cleaning and disinfecting needs.
  • Kills common microbiological contaminations on surfaces
  • Can be used in a variety of applications
  • Ideal for large surface cleaning
  • No strong odor provides good user acceptability
  • No special disposal is required
  • Facilitates transfer disinfection into a cleanroom
  • No mixing required
With broad-spectrum, best-in-class efficacy claims (30 kill claims), VersaQuat™ delivers consistent quality and intuitive usage. Recommended for use as a standard disinfectant as part of a disinfectant rotation program, VersaQuat™ is ideal for use in laboratories, hospitals, home healthcare settings, and veterinary clinics. It can also be used in offices, schools, gyms, and homes. Quick, effective, and convenient, VersaQuat™ is not abrasive and is compatible with various hard, non-porous surfaces.
Certifications: EPA registration number: 1839-220-92998. CA Dept. of Pesticides Regulation Number: 1839-220-AA-92998.