Puritan® Tongue Depressors, Puritan Medical Products
Catálogo # 62505-011
Proveedor: PURITAN MEDICAL PRODUCTS COMP
Product available on GSA Advantage®
Especificaciones
- Esterilidad:Sterile
- Anchura:19 mm (0.75")
- Descripción:Regular, individually wrapped
- Longitud:152 mm (6")
- Referencia:62505-011
- Nº de proveedor:25-705
Especificaciones
Acerca de este artículo
Smooth, splinter-free tongue blades are made from northern white birch. May be used as a clinical diagnostic aid, or as a stirrer for liquid medicine. Availble in sterile or nonsterile.
May be used as a clinical diagnostic aid, or as a stirrer for liquid medicine. Availble in sterile or nonsterile.