Coventry™ Wrapped Foam Swabs, ITW Chemtronics®
Catalog # 15145-612
Supplier: ITW-CHEMTRONICS,INC
Specifications
- Descripción:Coventry™ Foam Swab, Wood Handle
- Referencia:15145-612
- Head Width:3.1 mm (0.12")
- Head Length:27 mm (1.06")
- Handle Width:2.2 mm (0.09")
- Nº de proveedor:20050
Specifications
About this item
This durable adhesive-free swab are designed for precise applications and provides good particle entrapment and works well as a adhesive, lubricant and solvent applicators.
- Wicking action pulls solvent from surfaces
- Fiberless construction does not generate loose fibers or particles
- No adhesives or binders are used in the construction
- Stronger head than other foam swabs
Its small tip is ideal for use in tight tolerance areas. The swab has a wrapped, open-cell polyurethane foam head on a nylon or wood handle. Wrapped foam is durable and delivers superior abrasion resistance for the removal of tough contaminants.