GC Inlet Liners, SGE
Proveedor: Trajan Scientific and Medical
Certificados
Acerca de este artículo
This wide range of fully deactivated inlet liners is compatibile with the industry's most popular instrument makes and models. Liners cover a variety of applications, and ensure complete sample transfer from syringe to column without loss. The improved designs result in increased precision and accuracy of sample transfer.
Liners cover a variety of applications, and ensure complete sample transfer from syringe to column without loss. The improved designs result in increased precision and accuracy of sample transfer.
Varian® 1177 injectors accept all Agilent Technologies 6.3 mm O.D. liners.
Información relacionada
Frequently Bought Together