MiraSWAB® 1500 Series Swab, Foamtec International
89069-030
- Descripción:Semi-Flexible Small Bulb Tip
- Referencia:89069-030
- Longitud del mango:58.9 mm (2.315")
- Head Width:3.9 mm (0.153")
- Head Length:10.4 mm (0.409")
- Handle Width:2.5 mm (0.098")
- Nº de proveedor:HT1502FC-500
Swab features a unique microfiber fabric over a flexible foam tip thermally bonded to a polypropylene shaft
This design combines the cleaning performance of microfiber fabrics with the scratch-free properties of foam for optical, HCC, semiconductor, and life science applications. Excellent surface contact takes off hard-to-remove contaminants. Abrasion-resistant and compatible with many solvents.