ContactUsLinkComponent
SGE GC Column, BP1, Trajan Scientific and Medical
SGE GC Column, BP1, Trajan Scientific and Medical
Catalog # 60362-094
undefined
SGE GC Column, BP1, Trajan Scientific and Medical
Catalog # 60362-094
Supplier Number:  054096
Specifications
  • Formato de columna:
    Capillary
  • Espesor de la película:
    5.00 µm
  • Ø int.:
    0.53 mm
  • Longitud:
    50 m
  • Referencia:
    60362-094
  • Packing Material:
    Qc5/Bp1-5nse
  • Phase:
    BP1
  • Nº de proveedor:
    054096
Specifications
About this item
These non-polar general purpose columns are ideal for all types of routine analysis.
  • Low bleed
  • Classic dimethyl polysiloxane technology with high temperature cross-linking
  • Temperature range 0.1 to 1.0 µm film thickness: –60 to 340/360 °C (–76 to 644/680 °F); 1.5 to 3.0 µm film thickness: –60 to 300/320 °C (–76 to 572/608 °F); 4.0 to 5.0 µm film thickness: –60 to 280/300 °C (–76 to 536/572 °F)
Similar phases for BP1 are DB®-1, DB®-Petro, HP®-1, HP®-1ms, Rtx®-1, Ultra-1, SPB™-1, SPB-1 Sulfur, Petrocol™ DH, CP-Sil 5 CB, VB-1, ZB-1, and VF-1ms. Similar phases to BP1-PONA are Petrocol™ DH, and DB®-Petrol 100.