Specifications
- Descripción:Small Compressed Foam Swab
- Bond:Thermal
- Material del asa:Polypropylene
- Material de la cabeza:100 ppi Open-Cell CleanFoam®
- Color del mango:Light Green
- Referencia:TWTX751B
- Longitud del mango:50.0 mm (1.970")
- Longitud total:67.0 mm (2.638")
- Head Width:3.5 mm (0.140")
- Head Length:17.0 mm (0.670")
- Handle Width:2.5 mm (0.100")
- Nº de proveedor:TX751B
Specifications
About this item
The Small Compressed CleanFoam® Swab is an excellent swab for cleaning very small areas, especially thin grooved areas of parts and assemblies
The pointed, precision tip is especially useful for magnified or microscopic work since the tip will often obscure the whole field. An excellent swab for cleaning, it is suitable for precision application of solvents and cleaning solutions. Swab features a small compressed CleanFoam® head with pointed tip on a short rigid polypropylene handle. Its compressed, laundered CleanFoam® head possesses good firmness and a pointed tip. Polyurethane foam head has been solvent-washed to greatly reduce nonvolatile residue. A compact, rigid handle and tapered internal head core provide excellent support. The compact polypropylene handle provides support and control. Low particle generation and extractable levels. Excellent chemical resistance.