- Descripción:Heavy-Duty Support Stand
- Referencia:97035-520
- Dimensiones de la base:419×305 mm (16.5×12")
- Altura de la varilla:152.4 cm (60")
Heavy-duty support stands permit cylindrical vessels up to 30.5 cm (12") in diameter to sit close to the support rod.
- Cast iron base with black enamel finish
- Optional stainless steel support rods
- Rubber cushions under each support stand corner
Ideal for mounting stirring motors for heavy equipment
Accessories include extra stainless steel support rods and two choices of clamps. The support stand clamp (58950-660) is designed with a precision fit to accept a 13mm (1/2") diameter support shaft and a 16mm (5/8") diameter support rod on a support stand. The heavy-duty support clamp (14010-000) is made of a strong aluminum alloy and is fitted with oversize knobs for very secure positioning. Accepts shafts 6–24mm (0.24–0.94") in diameter and supports rods on a 10.2 cm (4") long surface to avoid wobble and vibration.
Base is constructed of cast iron with a black enamel finish. Each of the four corners rests on a rubber cushion. Includes a 16 mm (5/8") diameter support rod.