Absorbent Liners, Jaece
50.8 cm x 15 m (20" x 50') One Roll in Dispenser Box
Catalog # 10159-110
Supplier: JAECE INDUSTRIES INC
Supplier #: AL20501
Jump to:
Product Details & Documents
Keeps spills from spreading and protects benchtops, floors, and carts. Cushions surfaces while absorbing liquids. Made of highly absorbent three-ply tissue with a waterproof polypropylene backing that is resistant to most chemicals. Can be cut to size.
- Cushions surfaces while absorbing liquids
- Can be cut to size
Keeps spills from spreading and protects benchtops, floors, and carts.
Ordering information: Liner 82002-606 is packaged one roll per case.
Specifications
- Description:50.8 cm x 15 m (20" x 50') One Roll in Dispenser Box