ContactUsLinkComponent
VWR® Neoprene Flask Underlay
VWR® Neoprene Flask Underlay
Catalog # 56255-006
Supplier:  VWR International
undefined
VWR® Neoprene Flask Underlay
Catalog # 56255-006
Supplier:  VWR International
Specifications
  • Ø:
    13 cm (51/8")
  • Referencia:
    56255-006
Specifications
About this item
Moderate heat and oil resistance with concave recess on top to support round-bottom flasks and dishes.
  • Made of long-lasting green Neoprene rubber
Holds 250 and 500 mL flasks firmly in place.