Orders LinkContactUsLinkComponent
VWR® Ring Supports, Laboratory
VWR® Ring Supports, Laboratory
   Opciones sostenibles disponibles
Supplier:   VWR International
VWR® Ring Supports, Laboratory
Supplier:  VWR International
   Opciones sostenibles disponibles
Certificates
VWR® Ring Supports, Laboratory
4 Options Available Below
Product Details & Documents
Ring supports are made of compressed cork for supporting flasks or dishes.
  • Compressed cork construction
  • Internal bevel at top
  • Solid support for round-bottomed vessels
An internal bevel at top provides a secure support for round-bottomed vessels.
Specifications for Product Family
Especificaciones
  • Catalog No:
  • 56250-024
  • 56250-046
  • 56250-068
  • 56250-104
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
Especificaciones
Product Family Options
Información del producto
  • Ext.Ø
    For Flask Size
    Inner Diameter
    Availability
    Price
  • 80 mm (3")
    10 - 100 ml
    32 mm (1¼")
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 3
    Paquete de 3 - $214.01
    $214.01
    Paquete de 3
       $214.01
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    56250-024
    Environmentally Preferable
    Low Manufacturing Impact
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 110 mm (4")
    200 - 500 ml
    60 mm (2²⁄₅")
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 3
    Paquete de 3 - $368.71
    $368.71
    Paquete de 3
       $368.71
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    56250-046
    Environmentally Preferable
    Low Manufacturing Impact
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 140 mm (51/2")
    1000 - 3000 ml
    90 mm (3½")
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 3
    Paquete de 3 - $496.92
    $496.92
    Paquete de 3
       $496.92
    Especificaciones:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    56250-068
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • 210 mm (81/4")
    12 L
    150 mm (6")
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 3
    Paquete de 3 - $1,110.39
    $1,110.39
    Paquete de 3
       $1,110.39
    Especificaciones:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    56250-104
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact