Puritan® Glue Free Cotton Tipped Applicator, Wooden Handle, Puritan Medical Products
Catalog # 10806-005
Supplier: PURITAN MEDICAL PRODUCTS COMP
Product available on GSA Advantage®
Specifications
- Referencia:10806-005
- Longitud del mango:152.4 mm (6.000")
- Head Width:4.70 mm (0.188")
- Head Length:15.90 mm (0.625")
- Handle Width:2.5 mm (0.100")
- Nº de proveedor:867-WC NO GLUE
Specifications
About this item
Swab features a special construction that eliminates chemical migration.
Wood shaft. Adhesive-free.
Certifications: CE marked.
Ordering information: Packaged 100 swabs per bag, 10 bags per box, 10 boxes per case.