Puritan® Double Cotton Tipped Applicator, Puritan Medical Products
Catalog # 82030-746
Supplier: PURITAN MEDICAL PRODUCTS COMP
Product available on GSA Advantage®
Specifications
- Descripción:Small Tip, Fine Paper Handle
- Referencia:82030-746
- Longitud del mango:76.2 mm (3.000")
- Head Width:2 mm (0.078")
- Head Length:8.0 mm (0.314")
- Handle Width:1.5 mm (0.059")
- Nº de proveedor:890-PC DBL
Specifications
About this item
For use in cleaning sensitive components
Swabs feature a white, fine paper shaft, with fine, slim double-end cotton tips. Lint-free and highly absorbent. Antistatic packaging. Made in USA.