VWR® Critical Swab® Extra Large Cotton Tipped Swab, Wooden Handle
89031-272
Product available on GSA Advantage®
- Referencia:89031-272
- Longitud del mango:152.0 mm (6")
- Head Width:5.9 mm (0.234")
- Head Length:17.4 mm (0.687")
- Handle Width:2.5 mm (0.1")
Disposable swab with large absorbent cotton tip and wood shaft.
- Autoclavable
- Disposable
- Wood shaft
: CE marked.
: Packaged 100 swabs per bag, 5 bags per box, 10 boxes per case.