Orders LinkContactUsLinkComponent
Rtx®-624 GC Columns, Restek
Rtx®-624 GC Columns, Restek
Rtx-624, Capillary, 1.80 µm, 0.32 mm, 30 m
Catalog # RK10970
Supplier:   RESTEK CORP
Número de proveedor #:  10970
Rtx®-624 GC Columns, Restek
Catalog # RK10970
Supplier:  RESTEK CORP
Rtx®-624 GC Columns, Restek
Rtx®-624 GC Columns, Restek
Rtx-624, Capillary, 1.80 µm, 0.32 mm, 30 m
Price
$31,071.36
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
These gas chromatography columns are designed for the analysis of volatile organic compounds using a GC/MS system.
  • Well-suited for EPA method 524.2
  • Application-specific columns for volatile organic pollutants
  • Rtx®-624 phase produces greater than 90% resolution of the first six gases in EPA methods 8260 and 524.2
Equivalent to USP G43 phase
Phase composition: crossbonded 6% cyanopropylphenyl, 94% dimethylpolysiloxane. Temperature range: −20 to 240 °C (−4 to 464 °F). Recommended for EPA methods 502.2, 524.2, 601, 602, 624, 8010, 8020, 8240, and 8260.
Application-specific columns for volatile organic pollutants. The Rtx®-624 phase produces greater than 90% resolution of the first six gases in EPA methods 8260 and 524.2. This stationary phase is especially well-suited for EPA method 524.2 since it resolves 2-nitropropane from 1,1-dichloropropanone, which share quantification ion m/z 43 and must be separated chromatographically.
Especificaciones
  • Column Format:
    Capillary
  • Film thickness:
    1.80 µm
  • Ø int.:
    0.32 mm
  • Description:
    Rtx-624
  • Length:
    30 m