Orders LinkContactUsLinkComponent
Rtx®-5 Amine Columns, Restek
Rtx-5 Amine Column, Capillary, 1.00 µm, 0.32 mm, 30 m
Catalog # RK12354
Supplier:   RESTEK CORP
 :  12354
Rtx®-5 Amine Columns, Restek
Catalog # RK12354
Supplier:  RESTEK CORP
Rtx®-5 Amine Columns, Restek
Rtx®-5 Amine Columns, Restek
Rtx-5 Amine Column, Capillary, 1.00 µm, 0.32 mm, 30 m
Price
$32,724.24
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
These base-deactivated, low-polarity columns are designed for the analysis of basic compounds such as alkylamines, diamines, triamines, ethanolamines, and nitrogen-containing heterocycles, as well as neutral compounds, adsorptive compounds with oxygen groups susceptible to hydrogen bonding, and mildly acidic compounds such as phenol.
  • Eliminate the need to derivatize basic compounds
  • Low-polarity column
  • Internal dimensions and film thickness vary between models
Rtx®-5 Amine columns eliminate the need to derivatize the basic compounds before analysis while reducing adsorption and improving response for these compounds. Phase composition: Crossbond® 5% phenyl, 95% dimethylpolysiloxane.
Especificaciones
  • Column Format:
    Capillary
  • Film thickness:
    1.00 µm
  • Ø int.:
    0.32 mm
  • Description:
    Rtx-5 Amine Column
  • Length:
    30 m
  • Cat. No.:
    RK12354
  • Temperature limit:
    –60 to 300/315°C