SP Bel-Art Rotary Pipette Stand, Bel-Art Products, a part of SP
Catálogo # 53577-137
Proveedor: Bel-Art Products, a Part of SP
Especificaciones
- Descripción:Rotary Pipette Stand
- Referencia:53577-137
- Nº de proveedor:189570000
Especificaciones
Acerca de este artículo
Unique polypropylene and PVC stand holds up to 94 pipettes and rotates on a central vertical axis for convenient pipette selection.
- Polypropylene and PVC stand
- Convenient pipette selection with central vertical axis of rotation
- Provided with instructions for quick assembly
The lower rotating disc has four concentric troughs with tapered sides and open bottoms to protect tips and allow drainage. The upper rotating disc has a grid pattern concentric with the bottom, which keeps the pipettes vertical. The outer rim of the stationary base retains any draining liquid. A low center of gravity stabilizes the stand. It comes unassembled with instructions for quick assembly on the base.