ContactUsLinkComponent
SP Bel-Art Bacterial Cell Spreaders, Bel-Art Products, a part of SP
SP Bel-Art Bacterial Cell Spreaders, Bel-Art Products, a part of SP
Catálogo # 53800-008
undefined
SP Bel-Art Bacterial Cell Spreaders, Bel-Art Products, a part of SP
Catálogo # 53800-008
Número de proveedor:  F37736-0009
Especificaciones
  • Descripción:
    Large
  • Referencia:
    53800-008
  • Blade Width:
    25.4 mm (1")
  • Longitud del mango:
    20.3 cm (8")
  • Nº de proveedor:
    377360009
Especificaciones
Acerca de este artículo
For spreading cells onto the surface of an agar plate to form individual colonies.
  • Stainless steel construction
  • Autoclavable
  • May also be sterilized by flame
  • Smooth triangular end
These stainless spreaders have a smooth triangular end with a straight spreading bar. The small size is ideal for 6 cm plates and the large size is ideal for 9 cm plates. Spreaders are autoclavable and may be flame sterilized.