Orders LinkContactUsLinkComponent
VWR® Bacti Cell Spreaders
VWR® Bacti Cell Spreaders
VWR® Bacti Cell Spreaders
VWR® Bacti Cell Spreaders
VWR® Bacti Cell Spreaders
Convenient and economical cell spreaders are designed to simplify bacterial culture work.
  • Autoclavable
  • Simplify bacterial culture work
  • Durable fabrication
Durable enough to be autoclaved and reused.
Ordering information: Cell spreaders are available sterile (individually wrapped) or nonsterile (bulk packed).
Documents
Product Family Options
  • Color
    Overall Length
    Blade Width
    Sterility
  • Blue
    21.5 cm
    30 mm
    Non sterile
    Paquete de 25
    Paquete de 25 - $1,022.70
    $1,022.70
    Case de 4 (25UN)
    Case de 4 (25UN) - $1,114.74
    $1,114.74
    Paquete de 25
       $1,022.70
    Case de 4 (25UN)
       $1,114.74
  • Blue
    21.5 cm
    30 mm
    Sterile
    Paquete de 25
    Paquete de 25 - $2,361.97
    $2,361.97
    Case de 4 (25UN)
    Case de 4 (25UN) - $2,574.54
    $2,574.54
    Paquete de 25
       $2,361.97
    Case de 4 (25UN)
       $2,574.54
  • Yellow
    24.0 cm
    6.0 cm
    Non sterile
    Paquete de 25
    Paquete de 25 - $1,022.70
    $1,022.70
    Case de 4 (25UN)
    Case de 4 (25UN) - $1,114.74
    $1,114.74
    Paquete de 25
       $1,022.70
    Case de 4 (25UN)
       $1,114.74
  • Yellow
    24.0 cm
    6.0 cm
    Sterile
    Paquete de 25
    Paquete de 25 - $2,361.97
    $2,361.97
    Case de 4 (25UN)
    Case de 4 (25UN) - $2,574.54
    $2,574.54
    Paquete de 25
       $2,361.97
    Case de 4 (25UN)
       $2,574.54