Inoculating Loops and Needles, Sterile, Greiner Bio-One
Supplier: GREINER BIO-ONE
Certificates
About this item
Inoculating loops allow easier bacterial collection and inoculation.
- Color-coded
- Polystyrene construction
- Sterile
Inoculating needles are ideal for picking up single colonies.
Frequently Bought Together