ContactUsLinkComponent
L-Shaped Plastic Spreaders, Sterile
  30002-116
undefined
L-Shaped Plastic Spreaders, Sterile
  30002-116
 :  174CS01
  • Referencia:
    30002-116
  • Empaquetado:
    1/Peel Pack
  • Nº de proveedor:
    174CS01
Spreaders feature a completely smooth rounded surface, free of rough edges and imperfections in the plastic
This ensures that liquid samples are spread evenly across the surface of agar plates without gouging or cutting the medium.